IBC bunds, GRP
Toimittaja: CEMO
10398 10399 7588
CEMG10398EA 937 EUR
CEMG10398 CEMG10399 CEMG7588
IBC bunds, GRP
Imeytysaineet ja imeytysliinat
Practical storage and filling stations for water-polluting and flammable liquids. The IBC bunds are made from glass fibre reinforced plastic, GRP for intermediate bulk containers.
  • Flash point about 55 °C
  • Corrosion resistant
  • With general construction inspection approval
  • Capacity approx. 1000 L
These bunds are universally applicable for oil, lyes and acids.