Sump Pallets
Toimittaja: CEMO
10704 10705 10706 10707 10708 10709 10710 10711 10712 10713
CEMG10704EA 95.9 EUR
CEMG10704 CEMG10705 CEMG10706 CEMG10707 CEMG10708 CEMG10709 CEMG10710 CEMG10711 CEMG10712 CEMG10713
Sump Pallets
Tynnyrivarasto
These small container/laboratory trays can be combined to cover a complete storage area and also can be used as storage/filling station. Trays can be used with or without perforated plate.
  • Made of polyethylene
  • With general construction inspection approval (application for admission)
  • Stackable and provides space-saving storage/transport
  • Fully recyclable and maintenance friendly
  • Rugged, removable PE grating
  • Can be connected to form a surface protection system
Stable PE access ramp with an integrated fixing bolt for connection to the PE grating, slightMore Product Information