Erucic acid

Toimittaja: Cayman Chemical

90175-100 90175-250 90175-50 90175-500
CAYM90175-100EA 31.2 EUR
CAYM90175-100 CAYM90175-250 CAYM90175-50 CAYM90175-500
Erucic acid
Erucic acid
Kaava: C₂₂H₄₂O₂
MDL numero: MFCD00063188
CAS-numero: 112-86-7
EINECS: 204-011-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR