5,8,11,14-Eicosatetraynoic acid

Toimittaja: Cayman Chemical

90120-10 90120-5 90120-50 90120-25
CAYM90120-10EA 167 EUR
CAYM90120-10 CAYM90120-5 CAYM90120-50 CAYM90120-25
5,8,11,14-Eicosatetraynoic acid
5,8,11,14-Eicosatetraynoic acid
Kaava: C₂₀H₂₄O₂
MDL numero: MFCD00036967
CAS-numero: 1191-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR