(±)-Jasmonic acid

Toimittaja: Cayman Chemical

88300-1 88300-250 88300-500 88300-100
CAYM88300-1EA 363 EUR
CAYM88300-1 CAYM88300-250 CAYM88300-500 CAYM88300-100
(±)-Jasmonic acid
(±)-Jasmonic acid
Kaava: C₁₂H₁₈O₃
MDL numero: MFCD04221274
CAS-numero: 77026-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR