Griess reagent R2
Toimittaja: Cayman Chemical
780020-8 780020-50
CAYM780020-8EA 35.1 EUR
CAYM780020-8 CAYM780020-50
Griess reagent R2
Griess reagent
Griess Reagent component for determination of nitrite in aqueous solutions.