Calcitriol ≥97%, crystalline solid

Toimittaja: Cayman Chemical
Vaara

71820-5 71820-10 71820-1
CAYM71820-5EA 537 EUR
CAYM71820-5 CAYM71820-10 CAYM71820-1
Calcitriol ≥97%, crystalline solid
Calcitriol

Calcitriol is synthesised from 7-dehydro cholesterol in humans by a non-enzymatic photochemical reaction with 290 to 310 nm UV light in the skin.


The main physiologic effects of calcitriol are to increase the absorption of calcium at the level of the intestinal epithelium, and to increase the mineralization of bone via the direct stimulation of osteoblasts.

Kaava: C₂₇H₄₄O₃
MDL numero: MFCD00867079
CAS-numero: 32222-06-3
UN: 2811
ADR: 6.1,I

Order Now

Testitulosten erittely

Purity ≥97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR