N,N,N',N'-tetrametyyli-p-fenyyleenidiamiini-2HCl

Toimittaja: Cayman Chemical
Varoitus

Synonyms: N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

70455-5 70455-10 70455-25 70455-50
CAYM70455-5EA 59.8 EUR
CAYM70455-5 CAYM70455-10 CAYM70455-25 CAYM70455-50
N,N,N',N'-tetrametyyli-p-fenyyleenidiamiini-2HCl
N,N,N',N'-tetrametyyli-p-fenyyleenidiamiini-2HCl
Kaava: C₁₀H₁₈Cl₂N₂
Sulamispiste: 227…230 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012482
CAS-numero: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e955f6b2a1434ddba239f533c6611583219f8904&vid=13fb110b013db70c026f8eefeee7f08fa2568c0c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR