3,3',5,5'-Tetramethylbenzidine dihydrochloride monohydrate

Toimittaja: Cayman Chemical

70450-250 70450-1 70450-5
CAYM70450-250EA 94.9 EUR
CAYM70450-250 CAYM70450-1 CAYM70450-5
3,3',5,5'-Tetramethylbenzidine dihydrochloride monohydrate
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Kaava: C₁₆H₂₀N₂·2HCl·xH₂O
MDL numero: MFCD00150104
CAS-numero: 207738-08-7
EINECS: 264-769-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR