tetranor-PGDM ELISA kit
Toimittaja: Cayman Chemical
501001-96S 501001-96 501001-480S 501001-480
CAYM501001-96SEA 752 EUR
CAYM501001-96S CAYM501001-96 CAYM501001-480S CAYM501001-480
tetranor-PGDM ELISA kit
Bio-assayt ELISA-kitit
A competitive assay for the quantification of tetranor-PGDM in urine.
  • Detect tetranor-PGDM, a major metabolite of PGD₂ in urine
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure tetranor-PGDM levels down to 40 pg/ml
  • Read: Colorimetric at 405 to 420 nm
tetranor-PGDM is a major metabolite of prostaglandin D2 (PGD₂) found in human and mouse urine. In human urine, tetranor-PGDM is significantly more abundant than the PGD2 metabolites 11β-PGF2α and 2,3-dinor-11β-PGF₂α and is the only endogenous PGD₂ metabolite detectable in mouse urine by LC-MS.
Normal levels of tetranor-PGDM in human and mouse urine are 1.5 ng/mg creatinine and 8.1 ng/mg creatinine respectively. tetranor-PGDM ELISA Kit is a competitive assay that can be used for quantification of tetranor-PGDM in urine. This assay converts tetranor-PGDM to a stable derivative, tetranor-PGJM, that can be easily quantified.
Pakkaus: Kit contains: Antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.