Prostaglandin D₂-MOX Express ELISA kit
Toimittaja: Cayman Chemical
500151-96 500151-96S 500151-480S
CAYM500151-96EA 544 EUR
CAYM500151-96 CAYM500151-96S CAYM500151-480S
Prostaglandin D₂-MOX Express ELISA kit
Bio-assayt ELISA-kitit
A competitive assay that permits the rapid measurement of PGD₂ from biological samples.
  • Treatment of samples with MOX hydrochloride converts unstable PGD₂ to stable PGD₂-MOX, MOX derivatization reagent included with assay
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGD₂-MOX levels down to 16 pg/ml
  • Rapid assay; get results in under four hours
PGD₂-MOX Express ELISA is a competitive assay that permits the rapid measurement of PGD₂ from biological samples, requiring only one-hour incubation and development times for each step. The methodology for the assay is the same as that employed in the PGD₂-MOX ELISA (Item No. 512011). The ELISA offers the convenience of a fast assay while still achieving a sensitivity of ~16 pg/ml and an assay range of 7,8 to 1000 pg/ml.
Toimitustiedot: Kit contains: Antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, control, methoxylamine, sodium acetate, tracer and antiserum dye.