Tenofovir disoproxil fumarate ≥98%, crystalline solid

Toimittaja: Cayman Chemical
Vaara

16922-10 16922-25 16922-5 16922-50
CAYM16922-10EA 189 EUR
CAYM16922-10 CAYM16922-25 CAYM16922-5 CAYM16922-50
Tenofovir disoproxil fumarate ≥98%, crystalline solid
Tenofovir disoproxil fumarate

Tenofovir disoproxil is a prodrug of the acyclic nucleoside phosphonate, tenofovir. Tenofovir is converted by cellular enzymes to tenofovir diphosphate, an obligate chain terminator that inhibits the activity of HIV reverse transcriptase and hepatitis B virus polymerase. Due to its rapid intracellular uptake, the anti-HIV activity of tenofovir disoproxil is reportedly >100-fold greater than that of the negatively charged tenofovir in a T cell line and primary blood lymphocytes.


  • Crystalline solid
  • Soluble in organic solvents such as ethanol, DMSO and dimethyl formamide (DMF)
  • Sparingly soluble in aqueous buffers

Kaava: C₁₉H₃₀N₅O₁₀P·C₄H₄O₄
Storage Temperature: Pakastin
MDL numero: MFCD08141829
CAS-numero: 202138-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR