Apixaban ≥98%, crystalline solid

Toimittaja: Cayman Chemical
Vaara

15427-10 15427-5 15427-1 15427-50
CAYM15427-10EA 614 EUR
CAYM15427-10 CAYM15427-5 CAYM15427-1 CAYM15427-50
Apixaban ≥98%, crystalline solid
Apixaban

Apixaban is an orally bioavailable, selective inhibitor of both free and prothrombinase-bound factor Xa (Kis = 0.8 nM).


In vivo, apixaban has antithrombotic effects in a rabbit model of venous thrombosis. Apixaban (357.5 mg) also prevents thrombus formation without inducing adverse bleeding events in a porcine model of aortic heterotopic valve replacement. Formulations containing apixaban have been used to prevent blood clot formation in patients with atrial fibrillation.

Kaava: C₂₅H₂₅N₅O₄
Storage Temperature: Pakastin
MDL numero: MFCD11977295
CAS-numero: 503612-47-3

Order Now

Testitulosten erittely

Purity ≥98%
λmax (UV/Vis) 281 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR