Blasticidin S hydrochloride ≥95%, crystalline solid

Toimittaja: Cayman Chemical
Vaara

14499-10 14499-100 14499-25 14499-50
CAYM14499-10EA 68.9 EUR
CAYM14499-10 CAYM14499-100 CAYM14499-25 CAYM14499-50
Blasticidin S hydrochloride ≥95%, crystalline solid
Blasticidin S hydrochloride

Blasticidin S is a peptidyl nucleoside antibiotic. It was first isolated from S. griseochromogenes. The structure of the pentahydrate form is determined using diffractometer data and, similar to the antibiotic amicetin, the hydrophilic and hydrophobic groups lie on either side of the molecule.


  • Created using total synthesis
  • Stability: ≥2 years


Used in research to select mammalian cells transfected with the blasticidin resistance gene bsr.


Varoitus: Not for human or veterinary use.

Kaava: C₁₇H₂₇ClN₈O₅
Storage Temperature: Pakastin
MDL numero: MFCD02091640
CAS-numero: 3513-03-9
UN: 2811
ADR: 6.1,II

Order Now

Testitulosten erittely

Purity ≥95%
λmax 269 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR