Anti-TLR1 Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
13582-1
CAYM13582-1EA 725 EUR
CAYM13582-1
Anti-TLR1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TLR1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR