3-Bromo-5-phenylsalicylic acid

Toimittaja: Cayman Chemical

13574-1 13574-5 13574-10 13574-25
CAYM13574-1EA 114 EUR
CAYM13574-1 CAYM13574-5 CAYM13574-10 CAYM13574-25
3-Bromo-5-phenylsalicylic acid
3-Bromo-5-phenylsalicylic acid
Kaava: C₁₃H₉BrO₃
MDL numero: MFCD18378258
CAS-numero: 4906-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR