4-Nonylphenylboronic acid

Toimittaja: Cayman Chemical

13140-100 13140-250 13140-500 13140-1
CAYM13140-100EA 33.8 EUR
CAYM13140-100 CAYM13140-250 CAYM13140-500 CAYM13140-1
4-Nonylphenylboronic acid
4-Nonylphenylboronic acid
Kaava: C₁₅H₂₅BO₂
MDL numero: MFCD02093070
CAS-numero: 256383-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR