Ethyl palmitoleate

Toimittaja: Cayman Chemical

10008204-10 10008204-100 10008204-50 10008204-500
CAYM10008204-10EA 28.6 EUR
CAYM10008204-10 CAYM10008204-100 CAYM10008204-50 CAYM10008204-500
Ethyl palmitoleate
Ethyl palmitoleate
Kaava: C₁₈H₃₄O₂
MDL numero: MFCD00056314
CAS-numero: 56219-10-4
EINECS: 260-065-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR