Losartan potassium ≥98%, crystalline

Toimittaja: Cayman Chemical

10006594-10 10006594-100 10006594-50 10006594-500
CAYM10006594-10EA 29.9 EUR
CAYM10006594-10 CAYM10006594-100 CAYM10006594-50 CAYM10006594-500
Losartan potassium ≥98%, crystalline
Losartan potassium

Angiotensin II is a hormone that plays an important role in regulating blood pressure.


  • Synonyms: DuP-753; MK 954


Elevated levels of angiotensin II are implicated in inducing and maintaining hypertension, and also in the development of atherosclerosis. Both of these effects are mediated by the angiotensin II type 1 (AT1) receptor. Losartan is an AT1 receptor antagonist with a Ki value of 5 to 20 nM. It has an attenuating effect on vein graft atherosclerosis in rabbits and effectively reduces arterial blood pressure in rats. In humans, losartan controls hypertension while protecting renal function.

Kaava: C₂₂H₂₃ClN₆O
MDL numero: MFCD02092704
CAS-numero: 124750-99-8

Order Now

Testitulosten erittely

Purity ≥95%
Stability ≥2 years at −20 °C
Form A Crystaline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR