J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Columns, Recombinant Protein A Resin
Toimittaja: Avantor
C789-07EA 785 EUR
C789-07 C789-11 C789-18 C789-25
J.T.Baker® BAKERBOND® PROchievA™, Affinity Chromatography Columns, Recombinant Protein A Resin
Kromatografiakolonnit
The J.T.Baker® BAKERBOND® PROchievA™ recombinant protein A resin offers superior dynamic binding capacity with excellent alkaline stability linked to Avantor's proprietary ligand.
The new protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PROchievA™ recombinant protein A resin can be used with other J.T.Baker® BAKERBOND® resins toMore Product Information