Kalsiumklorididihydraatii ≥99.0%, Reagent Grade

Toimittaja: Honeywell Chemicals
Varoitus

C3881-500G C3881-12KG C3881-2.5KG
C3881-500GEA 32.99 EUR
C3881-500G C3881-12KG C3881-2.5KG
Kalsiumklorididihydraatii ≥99.0%, Reagent Grade
Kalsiumklorididihydraatii

  • ≥95% for general laboratory use.

Kaava: CaCl₂·2H₂O
Kiehumispiste: 1,900 °C (1.013 hPa)
Sulamispiste: 765 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149613
CAS-numero: 10035-04-8
EINECS: 233-140-8 

Order Now

Testitulosten erittely

Assay 99.0 - 105.0 %
Appearance of the solution complying
Colour of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR