Chlorazol black E, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY
Vaara

C1144-5G C1144-25G
C1144-5GEA 73.9 EUR
C1144-5G C1144-25G
Chlorazol black E, Sigma-Aldrich®
Chlorazol black E

Chlorazol Black has been used to monitor the infection of mycorrhizal Allium porrum in plant roots. It has also been used for the staining of copepods.

Kaava: C₃₄H₂₇N₉O₇S₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00066363
CAS-numero: 1937-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee571f233fb087fe2610096906b7c19399a6416b&vid=5af7869bad38a82ab38d5611cbd902b2b8313149&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR