Anti-CD95/FAS Mouse Monoclonal Antibody

Toimittaja: Bioworld Technology
MB0079
BWRLMB0079EA 376 EUR
BWRLMB0079
Anti-CD95/FAS Mouse Monoclonal Antibody
Vasta-aineet
Synthetic peptide, corresponding to the C-terminus of Human CD95/FAS.

This antibody detects endogenous levels of FAS (C-terminus) and does not cross-react with related proteins

Type: Primary
Antigen: CD95/FAS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR