Anti-HSP70 Mouse Monoclonal Antibody

Toimittaja: Bioworld Technology
MB0016
BWRLMB0016EA 376 EUR
BWRLMB0016
Anti-HSP70 Mouse Monoclonal Antibody
Vasta-aineet
Synthetic peptide, corresponding to the C-terminus of Human HSP70.

This antibody detects endogenous levels of HSPA1A and does not cross-react with related proteins

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR