Anti-WDR5 Rabbit Polyclonal Antibody

Toimittaja: Bioworld Technology
BS7354
BWRLBS7354EA 376 EUR
BWRLBS7354
Anti-WDR5 Rabbit Polyclonal Antibody
Vasta-aineet
Recombinant full length Human WDR5.

WDR5 pAb detects endogenous levels of WDR5 protein.

Type: Primary
Antigen: WDR5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR