Anti-HMBS Rabbit Polyclonal Antibody

Toimittaja: Bioworld Technology
BS6624
BWRLBS6624EA 376 EUR
BWRLBS6624
Anti-HMBS Rabbit Polyclonal Antibody
Vasta-aineet
Recombinant full length Human HMBS.

HMBS pAb detects endogenous levels of HMBS protein.

Type: Primary
Antigen: HMBS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR