Anti-p-ATM Rabbit Polyclonal Antibody [clone: S1987]

Toimittaja: Bioworld Technology
BS64020
BWRLBS64020EA 313 EUR
BWRLBS64020
Anti-p-ATM Rabbit Polyclonal Antibody [clone: S1987]
Vasta-aineet
A synthetic peptide corresponding to residues in Human ATM around the phosphorylation site of ser1987.

p-ATM (S1987) pAb detects endogenous levels of ATM protein only when phosphorylated at ser1987.

Type: Primary
Antigen: p-ATM
Clonality: Polyclonal
Clone: S1987
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR