Anti-Arrestin-beta-1 Rabbit Polyclonal Antibody

Toimittaja: Bioworld Technology
BS6102
BWRLBS6102EA 376 EUR
BWRLBS6102
Anti-Arrestin-beta-1 Rabbit Polyclonal Antibody
Vasta-aineet
Recombinant full length Human Arrestin-beta-1.

Arrestin-?-1 pAb detects endogenous levels of Arrestin-?-1 protein.

Type: Primary
Antigen: Arrestin-beta-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR