Anti-IL-1beta Rabbit Polyclonal Antibody

Toimittaja: Bioworld Technology
BS6067
BWRLBS6067EA 376 EUR
BWRLBS6067
Anti-IL-1beta Rabbit Polyclonal Antibody
Vasta-aineet
Recombinant full length Human IL-1beta.

IL-1? pAb detects endogenous levels of IL-1? protein.

Type: Primary
Antigen: IL-1beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR