Anti-Mitofilin Rabbit Polyclonal Antibody
Toimittaja: Bioworld Technology
BS60403
BWRLBS60403EA 289 EUR
BWRLBS60403
Anti-Mitofilin Rabbit Polyclonal Antibody
Vasta-aineet
A synthetic peptide corresponding to residues in Human Mitofilin.
Mitofilin pAb detects endogenous levels of Mitofilin protein.
Type: Primary
Antigen: Mitofilin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat