Anti-AMPKgamma1 Rabbit Polyclonal Antibody

Toimittaja: Bioworld Technology
BS5610
BWRLBS5610EA 289 EUR
BWRLBS5610
Anti-AMPKgamma1 Rabbit Polyclonal Antibody
Vasta-aineet
Synthetic peptide, corresponding to amino acids 36-81 of Human AMPKgamma1.

AMPK?1 pAb detects endogenous levels of AMPK?1 protein.

Type: Primary
Antigen: AMPKgamma1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR