Anti-p-SHP-1 Rabbit Polyclonal Antibody [clone: Y536]
Toimittaja: Bioworld Technology
BS4169
BWRLBS4169EA 313 EUR
BWRLBS4169
Anti-p-SHP-1 Rabbit Polyclonal Antibody [clone: Y536]
Vasta-aineet
Synthetic phosphopeptide derived from human SHP-1 around the phosphorylation site of Tyrosine 536.
p-SHP-1 (Y536) pAb detects endogenous levels of SHP-1 protein only when phosphorylated at Tyr536.
Type: Primary
Antigen: p-SHP-1
Clonality: Polyclonal
Clone: Y536
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat