Anti-p-CK18 Rabbit Polyclonal Antibody [clone: S33]

Toimittaja: Bioworld Technology
BS4113
BWRLBS4113EA 313 EUR
BWRLBS4113
Anti-p-CK18 Rabbit Polyclonal Antibody [clone: S33]
Vasta-aineet
Synthetic phosphopeptide derived from human Cytokeratin 18 around the phosphorylation site of Serine 33.

p-Cytokeratin 18 (S33) pAb detects endogenous levels of Cytokeratin 18 protein only when phosphorylated at Ser33.

Type: Primary
Antigen: p-CK18
Clonality: Polyclonal
Clone: S33
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR