Anti-Ang-1 Rabbit Polyclonal Antibody [clone: E207]

Toimittaja: Bioworld Technology
BS2829
BWRLBS2829EA 289 EUR
BWRLBS2829
Anti-Ang-1 Rabbit Polyclonal Antibody [clone: E207]
Vasta-aineet
Synthetic peptide, corresponding to amino acids 176-224 of Human Ang-1.

Ang-1 (E207) pAb detects endogenous levels of Ang-1 (E207) protein.

Type: Primary
Antigen: Ang-1
Clonality: Polyclonal
Clone: E207
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR