Anti-Frizzled-3 Rabbit Polyclonal Antibody [clone: L180]

Toimittaja: Bioworld Technology
BS2602
BWRLBS2602EA 289 EUR
BWRLBS2602
Anti-Frizzled-3 Rabbit Polyclonal Antibody [clone: L180]
Vasta-aineet
Synthetic peptide, corresponding to amino acids 150-200of Human Frizzled-3.

Frizzled-3 (L180) pAb detects endogenous levels of Frizzled-3 protein.

Type: Primary
Antigen: Frizzled-3
Clonality: Polyclonal
Clone: L180
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR