Anti-Matriptase Rabbit Polyclonal Antibody [clone: K21]

Toimittaja: Bioworld Technology
BS2574
BWRLBS2574EA 289 EUR
BWRLBS2574
Anti-Matriptase Rabbit Polyclonal Antibody [clone: K21]
Vasta-aineet
Synthetic peptide, corresponding to the N-terminual of Human Matriptase.

Matriptase (K21) pAb detects endogenous levels of Matriptase protein.

Type: Primary
Antigen: Matriptase
Clonality: Polyclonal
Clone: K21
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR