Anti-IKKalpha/beta Rabbit Polyclonal Antibody [clone: Q174]

Toimittaja: Bioworld Technology
BS1756
BWRLBS1756EA 289 EUR
BWRLBS1756
Anti-IKKalpha/beta Rabbit Polyclonal Antibody [clone: Q174]
Vasta-aineet
Synthetic peptide, corresponding to amino acids 151-200 of Human IKKalpha

IKK?/? (Q174) pAb detects endogenous levels of IKK?/? protein.

Type: Primary
Antigen: IKKalpha/beta
Clonality: Polyclonal
Clone: Q174
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR