Anti-Csk Rabbit Polyclonal Antibody [clone: L558]

Toimittaja: Bioworld Technology
BS1739
BWRLBS1739EA 289 EUR
BWRLBS1739
Anti-Csk Rabbit Polyclonal Antibody [clone: L558]
Vasta-aineet
Synthetic peptide, corresponding to amino acids C-terminus of Human Csk.

Csk (L558) pAb detects endogenous levels of Csk protein.

Type: Primary
Antigen: Csk
Clonality: Polyclonal
Clone: L558
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR