Anti-Paxillin Rabbit Polyclonal Antibody [clone: G112]

Toimittaja: Bioworld Technology
BS1288
BWRLBS1288EA 289 EUR
BWRLBS1288
Anti-Paxillin Rabbit Polyclonal Antibody [clone: G112]
Vasta-aineet
Synthetic peptide, corresponding to amino acids 100-150 of Human Paxillin.

Paxillin (G112) pAb detects endogenous levels of Paxillin protein.

Type: Primary
Antigen: Paxillin
Clonality: Polyclonal
Clone: G112
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR