Pipette container and basket
Toimittaja: BURKLE
9650-0653 9650-0652 9650-0651
BURK9650-0653EA 95.2 EUR
BURK9650-0653 BURK9650-0652 BURK9650-0651
Pipette container and basket
Korit Pipette Baskets
Made of PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
For cleaning and storage of pipettes. Containers and basket may be used as accessories for the pipette rinsing system.