Suppilot, rumpusuppilo
Toimittaja: BURKLE
9602-0350 9602-0250 9602-0400 9602-0450 9602-0200
BURK9602-0350EA 63 EUR
BURK9602-0350 BURK9602-0250 BURK9602-0400 BURK9602-0450 BURK9602-0200
Suppilot, rumpusuppilo
Suppilot Suodatussuppilot Tynnyrisuppilot
Made of PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.