Tubing, PTFE
Toimittaja: BURKLE
8879-0103 8804-0103 8804-0204 8804-0305 8804-0406 8804-0810 8804-1012 8879-0204 8879-1012
BURK8879-0103EA 275 EUR
BURK8879-0103 BURK8804-0103 BURK8804-0204 BURK8804-0305 BURK8804-0406 BURK8804-0810 BURK8804-1012 BURK8879-0204 BURK8879-1012
Tubing, PTFE
Letku Siirtoputki
PTFE, white, translucent.
  • Excellent chemical resistance
  • Very good thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
This tubing is characterised by its non stick surface and excellent dielectric properties.
Temperature resistance: −200 to +260 °C