Tubing, PVC
Toimittaja: BURKLE
8802-0612 8876-1016 8802-1218 8802-1016 8802-1320 8876-0612
BURK8802-0612EA 51.4 EUR
BURK8802-0612 BURK8876-1016 BURK8802-1218 BURK8802-1016 BURK8802-1320 BURK8876-0612
Tubing, PVC
Letku Siirtoputki
Pressure tubing made from reinforced fabric.
  • Permanent transparency
  • Increased flexibility
  • High chemical resistance
  • Excellent ageing and abrasion resistance
  • Environmentally friendly
  • Cadmium-free
Operating temperature: −20...+60 °C
Hardness (Shore durometer A): 70
Sertifikaatit: Foodstuffs quality according to the EC Guideline 1935/2004.
TÜV-approved.