Hose clips, worm-threaded
Toimittaja: BURKLE
8677-2540 8677-0812 8677-1016 8677-1220 8677-1625 8677-2032
BURK8677-2540EA 30.7 EUR
BURK8677-2540 BURK8677-0812 BURK8677-1016 BURK8677-1220 BURK8677-1625 BURK8677-2032
Hose clips, worm-threaded
Kourat Letkupuristimet
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.