Instrument trays, stainless steel
Toimittaja: BURKLE
4208-0001 4208-0002 4208-0003 4208-0004
BURK4208-0001EA 34.5 EUR
BURK4208-0001 BURK4208-0002 BURK4208-0003 BURK4208-0004
Instrument trays, stainless steel
Haihdutusmaljat/Tarjottimet/Altaat Instrumenttialustat/-astiat
Instruments, labware or samplers can be placed and transported on the sturdy stainless steel tray. The smooth surface is easy to clean.
  • Stainless steel V2A
  • Prepare, place and carry instruments and samples
  • Hygienic and robust
The rectangular instrument tray is suitable for hygienic placing in the fields of chemistry, pharmaceuticals, cosmetics, animal feed and food and beverage.