Pullo, kapea suuaukko, kierrekorkilla
Toimittaja: BURKLE
0302-3000 0302-0250 0302-0500 0302-1000 0302-0200 0302-0100 0302-5000
BURK0302-3000EA 28 EUR
BURK0302-3000 BURK0302-0250 BURK0302-0500 BURK0302-1000 BURK0302-0200 BURK0302-0100 BURK0302-5000
Pullo, kapea suuaukko, kierrekorkilla
Pullot Muovipullot
LDPE, translucent.
  • Cap absolutely tight closing
  • Suitable for use with foodstuffs
  • Good chemical and temperature resistance
Universal bottles for storing, packing, lab, taking samples, sampling and transport of liquids.