Parallel synthesis, Syncore® analyst
Toimittaja: BUCHI
1A30011N0 1A30011Y0 1A30012N0 1A30012Y0 1A30013N0 1A30013Y0 1A30021N0 1A30021Y0 1A30022N0 1A30022Y0 1A30023N0 1A30023Y0 1A30031N0 1A30031Y0 1A30032N0 1A30032Y0 1A30033N0 1A30033Y0 1A3C011N0 1A3C011Y0 1A3C012N0 1A3C012Y0 1A3C013N0 1A3C013Y0 1A3C021N0 1A3C021Y0 1A3C022N0 1A3C022Y0 1A3C023N0 1A3C023Y0 1A3C031N0 1A3C031Y0 1A3C032N0 1A3C032Y0 1A3C033N0 1A3C033Y0 1A3C211N0 1A3C211Y0 1A3C212N0 1A3C212Y0 1A3C213N0 1A3C213Y0 1A3C221N0 1A3C221Y0 1A3C222N0 1A3C222Y0 1A3C223N0 1A3C223Y0 1A3C231N0 1A3C231Y0 1A3C232N0 1A3C232Y0 1A3C233N0 1A3C233Y0 1A3S011N0 1A3S011Y0 1A3S012N0 1A3S012Y0 1A3S013N0 1A3S013Y0 1A3S021N0 1A3S021Y0 1A3S022N0 1A3S022Y0 1A3S023N0 1A3S023Y0 1A3S031N0 1A3S031Y0 1A3S032N0 1A3S032Y0 1A3S033N0 1A3S033Y0 1A3S211N0 1A3S211Y0 1A3S212N0 1A3S212Y0 1A3S213N0 1A3S213Y0 1A3S221N0 1A3S221Y0 1A3S222N0 1A3S222Y0 1A3S223N0 1A3S223Y0 1A3S231N0 1A3S231Y0 1A3S232N0 1A3S232Y0 1A3S233N0 1A3S233Y0
BUCH1A30011N0EA 0 EUR
BUCH1A30011N0 BUCH1A30011Y0 BUCH1A30012N0 BUCH1A30012Y0 BUCH1A30013N0 BUCH1A30013Y0 BUCH1A30021N0 BUCH1A30021Y0 BUCH1A30022N0 BUCH1A30022Y0 BUCH1A30023N0 BUCH1A30023Y0 BUCH1A30031N0 BUCH1A30031Y0 BUCH1A30032N0 BUCH1A30032Y0 BUCH1A30033N0 BUCH1A30033Y0 BUCH1A3C011N0 BUCH1A3C011Y0 BUCH1A3C012N0 BUCH1A3C012Y0 BUCH1A3C013N0 BUCH1A3C013Y0 BUCH1A3C021N0 BUCH1A3C021Y0 BUCH1A3C022N0 BUCH1A3C022Y0 BUCH1A3C023N0 BUCH1A3C023Y0 BUCH1A3C031N0 BUCH1A3C031Y0 BUCH1A3C032N0 BUCH1A3C032Y0 BUCH1A3C033N0 BUCH1A3C033Y0 BUCH1A3C211N0 BUCH1A3C211Y0 BUCH1A3C212N0 BUCH1A3C212Y0 BUCH1A3C213N0 BUCH1A3C213Y0 BUCH1A3C221N0 BUCH1A3C221Y0 BUCH1A3C222N0 BUCH1A3C222Y0 BUCH1A3C223N0 BUCH1A3C223Y0 BUCH1A3C231N0 BUCH1A3C231Y0 BUCH1A3C232N0 BUCH1A3C232Y0 BUCH1A3C233N0 BUCH1A3C233Y0 BUCH1A3S011N0 BUCH1A3S011Y0 BUCH1A3S012N0 BUCH1A3S012Y0 BUCH1A3S013N0 BUCH1A3S013Y0 BUCH1A3S021N0 BUCH1A3S021Y0 BUCH1A3S022N0 BUCH1A3S022Y0 BUCH1A3S023N0 BUCH1A3S023Y0 BUCH1A3S031N0 BUCH1A3S031Y0 BUCH1A3S032N0 BUCH1A3S032Y0 BUCH1A3S033N0 BUCH1A3S033Y0 BUCH1A3S211N0 BUCH1A3S211Y0 BUCH1A3S212N0 BUCH1A3S212Y0 BUCH1A3S213N0 BUCH1A3S213Y0 BUCH1A3S221N0 BUCH1A3S221Y0 BUCH1A3S222N0 BUCH1A3S222Y0 BUCH1A3S223N0 BUCH1A3S223Y0 BUCH1A3S231N0 BUCH1A3S231Y0 BUCH1A3S232N0 BUCH1A3S232Y0 BUCH1A3S233N0 BUCH1A3S233Y0
Parallel synthesis, Syncore® analyst
Haihduttimet Rinnakkaissynteesijärjestelmät
The Syncore® Analyst offers fast, gentle and environmentally friendly multiple sample concentration to a predefined volume.
  • Improve your sample throughput by concentrating up to 12 samples to a residual volume
  • No need of nitrogen saves operational cost
  • Optimize your process by combining the solid phase extraction and the concentration
  • Integrated solid phase extraction (SPE) eliminates contamination risks
  • High recovery rates due to smooth sample processing (up to 100 % solvent recovery)
Thanks to the orbital vortex movement, the water chilled appendix,More Product Information