Rotary evaporators, Rotavapor® R-300
Toimittaja: BUCHI
11SR300251VP11 11R300151V031 11R300151V131 11R300151V151 11R300151V051 11R300151V041 11R300151V141 11R300151V121 11R300151V021 11SR300111CS01 11SR300111VS01 11SR300112CS01 11SR300112VS01 11SR300151CS01 11SR300152CS01 11SR300211CS01 11SR300211VS01 11SR300212CS01 11SR300212VS01 11SR300251CS01 11SR300251VS02 11SR300251CP01 11SR300212VP01 11SR300212CP01 11SR300211VP01 11SR300211CP01 11SR300152VP01 11SR300111VS11 11SR300111CS11 11SR300112CS11 11SR300112VS11 11SR300151CS11 11SR300151VS11 11SR300152CS11 11SR300152VS11 11SR300211CS11 11SR300211VS11 11SR300212CS11 11SR300212VS11 11SR300251CS11 11SR300251VS11 11SR300252CS11 11SR300252VS11 11SR300151CP11 11SR300151VP11 11SR300251CP11 11SR300212VP11 11SR300212CP11 11SR300211CP11 11SR300152VP11 11SR300111VS21 11SR300111CS21 11SR300112CS21 11SR300112VS21 11SR300151CS21 11SR300151VS21 11SR300152CS21 11SR300152VS21 11R300251V141 11SR300211CS21 11SR300211VS21 11SR300212CS21 11SR300212VS21 11SR300251CS21 11SR300251VS21 11SR300252CS21 11SR300252VS21 11SR300151CP21 11SR300112VP21 11SR300112CP21 11SR300111VP21 11SR300111CP21 11SR300252VP21 11SR300252CP21 11SR300151VP21 11SR300251VP21 11SR300251CP21 11SR300212VP21 11SR300211VP21 11SR300211CP21 11SR300152VP21 11SR300152CP21 11R300151V001 11R300151V011 11R300151V101 11R300151V111 11R300201V111 11R300251V101 11SR300251VS01 11SR300111CP01 11SR300111VP01 11SR300112CP01 11SR300112VP01 11SR300151CP01 11SR300151VP01 11SR300252CP01 11SR300252CS01 11SR300252VP01 11SR300252VS01 11SR300111CP11 11SR300111VP11 11SR300112CP11 11SR300112VP11 11SR300252VP11
BUCH11SR300251VP11EA 16480 EUR
BUCH11SR300251VP11 BUCH11R300151V031 BUCH11R300151V131 BUCH11R300151V151 BUCH11R300151V051 BUCH11R300151V041 BUCH11R300151V141 BUCH11R300151V121 BUCH11R300151V021 BUCH11SR300111CS01 BUCH11SR300111VS01 BUCH11SR300112CS01 BUCH11SR300112VS01 BUCH11SR300151CS01 BUCH11SR300152CS01 BUCH11SR300211CS01 BUCH11SR300211VS01 BUCH11SR300212CS01 BUCH11SR300212VS01 BUCH11SR300251CS01 BUCH11SR300251VS02 BUCH11SR300251CP01 BUCH11SR300212VP01 BUCH11SR300212CP01 BUCH11SR300211VP01 BUCH11SR300211CP01 BUCH11SR300152VP01 BUCH11SR300111VS11 BUCH11SR300111CS11 BUCH11SR300112CS11 BUCH11SR300112VS11 BUCH11SR300151CS11 BUCH11SR300151VS11 BUCH11SR300152CS11 BUCH11SR300152VS11 BUCH11SR300211CS11 BUCH11SR300211VS11 BUCH11SR300212CS11 BUCH11SR300212VS11 BUCH11SR300251CS11 BUCH11SR300251VS11 BUCH11SR300252CS11 BUCH11SR300252VS11 BUCH11SR300151CP11 BUCH11SR300151VP11 BUCH11SR300251CP11 BUCH11SR300212VP11 BUCH11SR300212CP11 BUCH11SR300211CP11 BUCH11SR300152VP11 BUCH11SR300111VS21 BUCH11SR300111CS21 BUCH11SR300112CS21 BUCH11SR300112VS21 BUCH11SR300151CS21 BUCH11SR300151VS21 BUCH11SR300152CS21 BUCH11SR300152VS21 BUCH11R300251V141 BUCH11SR300211CS21 BUCH11SR300211VS21 BUCH11SR300212CS21 BUCH11SR300212VS21 BUCH11SR300251CS21 BUCH11SR300251VS21 BUCH11SR300252CS21 BUCH11SR300252VS21 BUCH11SR300151CP21 BUCH11SR300112VP21 BUCH11SR300112CP21 BUCH11SR300111VP21 BUCH11SR300111CP21 BUCH11SR300252VP21 BUCH11SR300252CP21 BUCH11SR300151VP21 BUCH11SR300251VP21 BUCH11SR300251CP21 BUCH11SR300212VP21 BUCH11SR300211VP21 BUCH11SR300211CP21 BUCH11SR300152VP21 BUCH11SR300152CP21 BUCH11R300151V001 BUCH11R300151V011 BUCH11R300151V101 BUCH11R300151V111 BUCH11R300201V111 BUCH11R300251V101 BUCH11SR300251VS01 BUCH11SR300111CP01 BUCH11SR300111VP01 BUCH11SR300112CP01 BUCH11SR300112VP01 BUCH11SR300151CP01 BUCH11SR300151VP01 BUCH11SR300252CP01 BUCH11SR300252CS01 BUCH11SR300252VP01 BUCH11SR300252VS01 BUCH11SR300111CP11 BUCH11SR300111VP11 BUCH11SR300112CP11 BUCH11SR300112VP11 BUCH11SR300252VP11
Rotary evaporators, Rotavapor® R-300
Haihduttimet Pyöröhaihduttimet
Its modular design allows the extension of the Rotavapor® R-300 to a fully integrated system where a central interface regulates each component.
  • Comfortable flask and vapor duct removal thanks to easy to use Combi-Clip
  • User-friendly operation due to ergonomic electronic or manual lift
  • Immediate adjustment of heating temperature and rotation speed with intuitively positioned control knobs
  • Easy plug & play extension to a fully integrated system including a central interface, vacuum pump and recirculating chiller
  • Precise operation control with integrated