Anti-WT1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: WT1/857]
Toimittaja: Biotium
BNUM0857-50
BTIUBNUM0857-50EA 417 EUR
BTIUBNUM0857-50
Anti-WT1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: WT1/857]
Vasta-aineet
Recognizes a 47-55 kDa-tumor suppressor protein, identified as Wilm's Tumor (WT1) protein. The antibody reacts with all isoforms of the full-length WT1 and also identifies WT1 lacking exon 2-encoded amino acids, frequently found in subsets of sporadic Wilm s tumors.WT1, a sporadic and familial pediatric kidney tumor, is genetically heterogeneous. Wilm s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, andMore Product Information