Anti-IGF1 Mouse Monoclonal Antibody (CF640R) [clone: IGF1/1020]
Toimittaja: Biotium
BNC401020-500 BNC401020-100
BTIUBNC401020-500EA 560 EUR
BTIUBNC401020-500 BTIUBNC401020-100
Anti-IGF1 Mouse Monoclonal Antibody (CF640R) [clone: IGF1/1020]
Vasta-aineet
This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin. IGF-1 is a polypeptide growth factor with two isoforms that are produced by alternative splicing. Isoform 1 is also known as IGF-IB while isoform 2 is known as IGF-IA. IGF-1 stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It functions as an autocrine regulator of growth. Activation of IGF system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like thoseMore Product Information